Product Details

SNP ID
rs12166503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41801266 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAATGTGGAGGGTCCTGGGTGAAC[C/T]TGGACTATTCCTTCTCTAGGTTTCA
Phenotype
MIM: 608797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC134 PubMed Links

Gene Details

Gene
CCDC134
Gene Name
coiled-coil domain containing 134
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304797.1 Intron NP_001291726.1
NM_024821.3 Intron NP_079097.1
XM_005261748.3 Intron XP_005261805.1
Gene
MEI1
Gene Name
meiotic double-stranded break formation protein 1
There are no transcripts associated with this gene.

View Full Product Details