Product Details
- SNP ID
-
rs11118070
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:218292591 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCTGTGAATGACTGCAAAAGTGCC[A/C]TGAGTATTGATTTGGGGTTACGAAT
- Phenotype
-
MIM: 611193
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RRP15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114625870] are located under a probe and SNP(s) [rs138022373] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RRP15
- Gene Name
- ribosomal RNA processing 15 homolog
View Full Product Details