Product Details

SNP ID
rs12085522
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:218290187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATCAATGAAAAAGAGTAACTGG[A/C]AACATGACCACATTGTTGGACATCA
Phenotype
MIM: 611193
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RRP15 PubMed Links

Gene Details

Gene
RRP15
Gene Name
ribosomal RNA processing 15 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016052.3 Intron NP_057136.2
XM_011509597.2 Intron XP_011507899.1

View Full Product Details