Product Details

SNP ID
rs7255784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12433757 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATTTTGTTACATACTCAGCAGAC[A/G]GAGTGCACTGATCAGCCACTGCTAG
Phenotype
MIM: 606697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF443 PubMed Links

Gene Details

Gene
ZNF443
Gene Name
zinc finger protein 443
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005815.4 1213 Intron NP_005806.2
XM_011527622.2 1213 Intron XP_011525924.1
XM_011527623.2 1213 Intron XP_011525925.2
XM_017026139.1 1213 Intron XP_016881628.1
XM_017026140.1 1213 UTR 5 XP_016881629.1

View Full Product Details