Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001324444.1 | 5182 | UTR 3 | NP_001311373.1 | ||
NM_001324445.1 | 5182 | UTR 3 | NP_001311374.1 | ||
NM_001324446.1 | 5182 | UTR 3 | NP_001311375.1 | ||
NM_001324448.1 | 5182 | UTR 3 | NP_001311377.1 | ||
NM_001324449.1 | 5182 | UTR 3 | NP_001311378.1 | ||
NM_001324450.1 | 5182 | UTR 3 | NP_001311379.1 | ||
NM_001324451.1 | 5182 | UTR 3 | NP_001311380.1 | ||
NM_001324452.1 | 5182 | UTR 3 | NP_001311381.1 | ||
NM_001324453.1 | 5182 | UTR 3 | NP_001311382.1 | ||
NM_012091.4 | 5182 | UTR 3 | NP_036223.2 |