Product Details

SNP ID
rs7456745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:48037652 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCAGGGAAGGCTATATATATATA[A/T]AAAAAATCTATAGGATAGTAAATGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C7orf57 PubMed Links

Gene Details

Gene
C7orf57
Gene Name
chromosome 7 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100159.2 Intron NP_001093629.1
NM_001267865.1 Intron NP_001254794.1
NM_001267866.1 Intron NP_001254795.1
XM_006715649.3 Intron XP_006715712.1
XM_006715650.2 Intron XP_006715713.1
XM_011515121.1 Intron XP_011513423.1
XM_011515122.2 Intron XP_011513424.1
XM_011515123.2 Intron XP_011513425.1
Gene
SUN3
Gene Name
Sad1 and UNC84 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details