Product Details

SNP ID
rs11024437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2887426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGATGTCACAATTTTCATCTTA[A/G]GAAAACAAAGAGAAGCTGAAGAAGG
Phenotype
MIM: 600856 MIM: 603240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDKN1C PubMed Links

Gene Details

Gene
CDKN1C
Gene Name
cyclin dependent kinase inhibitor 1C
There are no transcripts associated with this gene.

Gene
SLC22A18AS
Gene Name
solute carrier family 22 member 18 antisense
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302862.1 3639 UTR 3 NP_001289791.1
NM_007105.3 3639 UTR 3 NP_009036.2
XM_017017832.1 3639 UTR 3 XP_016873321.1
XM_017017833.1 3639 UTR 3 XP_016873322.1
XM_017017834.1 3639 UTR 3 XP_016873323.1
XM_017017835.1 3639 UTR 3 XP_016873324.1
XM_017017836.1 3639 UTR 3 XP_016873325.1

View Full Product Details