Product Details
- SNP ID
-
rs6657647
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:150740297 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAACAATTCTAAGTGAATGGGGTA[C/T]GAAGGAGATAGTATTTGAGGCAGGA
- Phenotype
-
MIM: 116845
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTSS
PubMed Links
Gene Details
- Gene
- CTSS
- Gene Name
- cathepsin S
View Full Product Details