Product Details

SNP ID
rs11084705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33081857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGGTTAGGGTTAGGGTCATGGG[A/G]AATTACACAGGGAGGGATATATATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH1 PubMed Links

Gene Details

Gene
GPATCH1
Gene Name
G-patch domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018025.2 Intron NP_060495.2
XM_006723255.3 Intron XP_006723318.1

View Full Product Details