Product Details

SNP ID
rs12930241
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30110854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGCATTCCAGCCCGAGCAAGA[A/C]TGTCTCAAAAAAAAAAAAAAAATTG
Phenotype
MIM: 616318 MIM: 601795
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GDPD3 PubMed Links
Additional Information
For this assay, SNP(s) [rs536906899] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GDPD3
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024307.2 Intron NP_077283.2
Gene
LOC101928595
Gene Name
uncharacterized LOC101928595
There are no transcripts associated with this gene.

Gene
MAPK3
Gene Name
mitogen-activated protein kinase 3
There are no transcripts associated with this gene.

View Full Product Details