Product Details

SNP ID
rs12049237
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:179300414 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGTTGGGCATCGTCTTGTGATTT[A/T]TTAGCCCTGACTTTTCCCTTAAGTT
Phenotype
MIM: 102642
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SOAT1 PubMed Links

Gene Details

Gene
SOAT1
Gene Name
sterol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252511.1 Intron NP_001239440.1
NM_001252512.1 Intron NP_001239441.1
NM_003101.5 Intron NP_003092.4
XM_011509911.1 Intron XP_011508213.1
XM_011509912.1 Intron XP_011508214.1

View Full Product Details