Product Details

SNP ID
rs12280580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:626220 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCAGCTGGGTGTGGTCAGTCCCT[C/G]CTGGGTGGATGGGCCTCCATCCATC
Phenotype
MIM: 606839 MIM: 182099
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDHR5 PubMed Links

Gene Details

Gene
CDHR5
Gene Name
cadherin related family member 5
There are no transcripts associated with this gene.

Gene
SCT
Gene Name
secretin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021920.3 670 Intron NP_068739.1
XM_017018109.1 670 UTR 3 XP_016873598.1

View Full Product Details