Product Details
- SNP ID
-
hCV31622220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:158935471 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAAATGAATTAGACAAATCAAGC[C/T]TAAAAGAAAAATCAAGCAACTATTG
- Phenotype
-
MIM: 612677
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PYHIN1
PubMed Links
Gene Details
- Gene
- PYHIN1
- Gene Name
- pyrin and HIN domain family member 1
There are no transcripts associated with this gene.
View Full Product Details