Product Details

SNP ID
rs11586024
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:152969457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTCCTATCATTTCTCAGGCCAT[C/T]CTACCCCTTCCTCTGGTGATGGCAG
Phenotype
MIM: 616363
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPRR4 PubMed Links

Gene Details

Gene
SPRR4
Gene Name
small proline rich protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173080.2 Intron NP_775103.1
XM_017000482.1 Intron XP_016855971.1

View Full Product Details