Product Details

SNP ID
rs12086457
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:110597538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGGTGCACACAGGAAGGAGACA[A/G]AGTGACAAAGCCCTCTCACAGGCCT
Phenotype
MIM: 176262
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNA2 PubMed Links

Gene Details

Gene
KCNA2
Gene Name
potassium voltage-gated channel subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204269.1 7949 Intron NP_001191198.1
NM_004974.3 7949 Intron NP_004965.1
XM_011541396.2 7949 UTR 3 XP_011539698.1
XM_011541397.2 7949 UTR 3 XP_011539699.1
XM_011541398.2 7949 UTR 3 XP_011539700.1
XM_011541399.2 7949 UTR 3 XP_011539701.1
XM_011541400.2 7949 UTR 3 XP_011539702.1
XM_017001213.1 7949 UTR 3 XP_016856702.1

View Full Product Details