Product Details
- SNP ID
-
rs11768472
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:50593065 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGAGAATTTGGTGTTCCCGTCAT[C/G]TAGGCTGAAGAACGTCTGCCCGTCG
- Phenotype
-
MIM: 601523
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GRB10
PubMed Links
Gene Details
- Gene
- GRB10
- Gene Name
- growth factor receptor bound protein 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001549.2 |
1975 |
Missense Mutation |
CAT,GAT |
H512D |
NP_001001549.1 |
NM_001001550.2 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
NP_001001550.1 |
NM_001001555.2 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
NP_001001555.1 |
NM_005311.4 |
1975 |
Missense Mutation |
CAT,GAT |
H558D |
NP_005302.3 |
XM_011515304.1 |
1975 |
Missense Mutation |
CAT,GAT |
H558D |
XP_011513606.1 |
XM_011515312.2 |
1975 |
Missense Mutation |
CAT,GAT |
H552D |
XP_011513614.1 |
XM_011515316.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_011513618.1 |
XM_011515323.2 |
1975 |
Missense Mutation |
CAT,GAT |
H497D |
XP_011513625.1 |
XM_017012029.1 |
1975 |
Missense Mutation |
CAT,GAT |
H674D |
XP_016867518.1 |
XM_017012030.1 |
1975 |
Missense Mutation |
CAT,GAT |
H652D |
XP_016867519.1 |
XM_017012031.1 |
1975 |
Missense Mutation |
CAT,GAT |
H629D |
XP_016867520.1 |
XM_017012032.1 |
1975 |
Missense Mutation |
CAT,GAT |
H603D |
XP_016867521.1 |
XM_017012033.1 |
1975 |
Missense Mutation |
CAT,GAT |
H603D |
XP_016867522.1 |
XM_017012034.1 |
1975 |
Missense Mutation |
CAT,GAT |
H603D |
XP_016867523.1 |
XM_017012035.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867524.1 |
XM_017012036.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867525.1 |
XM_017012037.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867526.1 |
XM_017012038.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867527.1 |
XM_017012039.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867528.1 |
XM_017012040.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867529.1 |
XM_017012041.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867530.1 |
XM_017012042.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867531.1 |
XM_017012043.1 |
1975 |
Missense Mutation |
CAT,GAT |
H597D |
XP_016867532.1 |
XM_017012044.1 |
1975 |
Missense Mutation |
CAT,GAT |
H596D |
XP_016867533.1 |
XM_017012045.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_016867534.1 |
XM_017012046.1 |
1975 |
Missense Mutation |
CAT,GAT |
H558D |
XP_016867535.1 |
XM_017012047.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867536.1 |
XM_017012048.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867537.1 |
XM_017012049.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867538.1 |
XM_017012050.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867539.1 |
XM_017012051.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867540.1 |
XM_017012052.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867541.1 |
XM_017012053.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867542.1 |
XM_017012054.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867543.1 |
XM_017012055.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867544.1 |
XM_017012056.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867545.1 |
XM_017012057.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867546.1 |
XM_017012058.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867547.1 |
XM_017012059.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867548.1 |
XM_017012060.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867549.1 |
XM_017012061.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867550.1 |
XM_017012062.1 |
1975 |
Missense Mutation |
CAT,GAT |
H545D |
XP_016867551.1 |
XM_017012063.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_016867552.1 |
XM_017012064.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_016867553.1 |
XM_017012065.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_016867554.1 |
XM_017012066.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_016867555.1 |
XM_017012067.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_016867556.1 |
XM_017012068.1 |
1975 |
Missense Mutation |
CAT,GAT |
H500D |
XP_016867557.1 |
View Full Product Details