Product Details

SNP ID
rs11188247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95241720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCAAAAGCACTAAGGGCACTTG[C/T]GTGGTTTGGCCCTGTTCCCACAGAG
Phenotype
MIM: 605900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDLIM1 PubMed Links

Gene Details

Gene
PDLIM1
Gene Name
PDZ and LIM domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020992.3 Intron NP_066272.1
XM_011540330.1 Intron XP_011538632.1

View Full Product Details