Product Details

SNP ID
rs12412369
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95607174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGTGTGTGGACTCTTCTTCTG[C/T]ATTTTATCCCCAGTGCATTTTGACA
Phenotype
MIM: 138250
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ALDH18A1 PubMed Links

Gene Details

Gene
ALDH18A1
Gene Name
aldehyde dehydrogenase 18 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017423.1 Intron NP_001017423.1
NM_001323412.1 Intron NP_001310341.1
NM_001323413.1 Intron NP_001310342.1
NM_001323414.1 Intron NP_001310343.1
NM_001323415.1 Intron NP_001310344.1
NM_001323416.1 Intron NP_001310345.1
NM_001323417.1 Intron NP_001310346.1
NM_001323418.1 Intron NP_001310347.1
NM_001323419.1 Intron NP_001310348.1
NM_002860.3 Intron NP_002851.2

View Full Product Details