Product Details

SNP ID
rs12279899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56177025 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCCTTGCTTTACACTGTAGCCAT[C/G]TCTGATAGAAAGTGTGTGGAGCTTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5J2 PubMed Links

Gene Details

Gene
OR5J2
Gene Name
olfactory receptor family 5 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005492.1 408 Missense Mutation ATC,ATG I136M NP_001005492.1

View Full Product Details