Product Details

SNP ID
rs3181263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67160978 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGAAGCACCCGGGACATCTCCCC[G/A]GCGAGGCGGAGGCGGGGAAGGAGCC
Phenotype
MIM: 609077 MIM: 602438 MIM: 605235 MIM: 603500
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
B3GNT9 PubMed Links

Gene Details

Gene
B3GNT9
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9
There are no transcripts associated with this gene.

Gene
FBXL8
Gene Name
F-box and leucine rich repeat protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018378.2 Intron NP_060848.2
Gene
HSF4
Gene Name
heat shock transcription factor 4
There are no transcripts associated with this gene.

Gene
NOL3
Gene Name
nucleolar protein 3
There are no transcripts associated with this gene.

Gene
TRADD
Gene Name
TNFRSF1A associated via death domain
There are no transcripts associated with this gene.

View Full Product Details