Product Details
- SNP ID
-
hCV31767485
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:4374845 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACCCCCAGGCCCAGGAAGCTTA[A/G]GGCTGGCGGCCTTCCTTGGTGACTA
- Phenotype
-
MIM: 611668
MIM: 608843
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CORO7
PubMed Links
Gene Details
- Gene
- CORO7
- Gene Name
- coronin 7
There are no transcripts associated with this gene.
- Gene
- CORO7-PAM16
- Gene Name
- CORO7-PAM16 readthrough
There are no transcripts associated with this gene.
- Gene
- VASN
- Gene Name
- vasorin
There are no transcripts associated with this gene.
View Full Product Details