Product Details

SNP ID
rs12258829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:35131924 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCTTCAAAAGTTTAAAAATTTG[A/G]TTTCACATTGGCCGGGTGCGGTGGC
Phenotype
MIM: 123812 MIM: 603135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREM PubMed Links

Gene Details

Gene
CREM
Gene Name
cAMP responsive element modulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267562.1 Intron NP_001254491.1
NM_001267563.1 Intron NP_001254492.1
NM_001267564.1 Intron NP_001254493.1
NM_001267565.1 Intron NP_001254494.1
NM_001267566.1 Intron NP_001254495.1
NM_001267567.1 Intron NP_001254496.1
NM_001267568.1 Intron NP_001254497.1
NM_001267569.1 Intron NP_001254498.1
NM_001267570.1 Intron NP_001254499.1
NM_001881.3 Intron NP_001872.3
NM_181571.2 Intron NP_853549.1
NM_182717.1 Intron NP_874386.1
NM_182718.1 Intron NP_874387.1
NM_182719.1 Intron NP_874388.1
NM_182720.1 Intron NP_874389.1
NM_182721.1 Intron NP_874390.1
NM_182723.1 Intron NP_874392.1
NM_182724.1 Intron NP_874393.1
NM_182769.2 Intron NP_877570.1
NM_182770.2 Intron NP_877571.1
NM_182771.1 Intron NP_877572.1
NM_182772.1 Intron NP_877573.1
NM_183011.1 Intron NP_898829.1
NM_183012.1 Intron NP_898830.1
NM_183013.2 Intron NP_898831.1
NM_183060.2 Intron NP_898883.1
XM_006717378.3 Intron XP_006717441.1
XM_006717379.3 Intron XP_006717442.1
XM_006717382.3 Intron XP_006717445.1
XM_006717383.3 Intron XP_006717446.1
XM_006717387.3 Intron XP_006717450.1
XM_011519316.2 Intron XP_011517618.1
XM_011519324.2 Intron XP_011517626.1
XM_011519325.2 Intron XP_011517627.1
XM_011519327.2 Intron XP_011517629.1
XM_011519330.2 Intron XP_011517632.1
XM_011519331.2 Intron XP_011517633.1
XM_011519332.2 Intron XP_011517634.1
XM_011519333.1 Intron XP_011517635.1
XM_011519335.2 Intron XP_011517637.1
XM_017015716.1 Intron XP_016871205.1
XM_017015717.1 Intron XP_016871206.1
XM_017015718.1 Intron XP_016871207.1
XM_017015719.1 Intron XP_016871208.1
XM_017015720.1 Intron XP_016871209.1
XM_017015721.1 Intron XP_016871210.1
XM_017015722.1 Intron XP_016871211.1
XM_017015723.1 Intron XP_016871212.1
XM_017015724.1 Intron XP_016871213.1
XM_017015725.1 Intron XP_016871214.1
XM_017015726.1 Intron XP_016871215.1
XM_017015727.1 Intron XP_016871216.1
XM_017015728.1 Intron XP_016871217.1
XM_017015729.1 Intron XP_016871218.1
XM_017015730.1 Intron XP_016871219.1
XM_017015731.1 Intron XP_016871220.1
XM_017015732.1 Intron XP_016871221.1
XM_017015733.1 Intron XP_016871222.1
XM_017015734.1 Intron XP_016871223.1
XM_017015735.1 Intron XP_016871224.1
XM_017015736.1 Intron XP_016871225.1
XM_017015737.1 Intron XP_016871226.1
Gene
CUL2
Gene Name
cullin 2
There are no transcripts associated with this gene.

View Full Product Details