Product Details

SNP ID
rs7398658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4806967 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAATAATGACAGATAACTGGAT[A/G]AGGGTAAAGTGACAGATAACTGGAT
Phenotype
MIM: 176257
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNA6 PubMed Links

Gene Details

Gene
KCNA6
Gene Name
potassium voltage-gated channel subfamily A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002235.3 91 Intron NP_002226.1
XM_005253686.3 91 UTR 5 XP_005253743.1
XM_011520955.2 91 UTR 5 XP_011519257.1
XM_017019270.1 91 UTR 5 XP_016874759.1
XM_017019271.1 91 UTR 5 XP_016874760.1
XM_017019272.1 91 UTR 5 XP_016874761.1

View Full Product Details