Product Details

SNP ID
rs4871340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123044674 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAATCAGAGCAACGGTAGGCCAGG[C/T]ATGTCTGGTCTTTTCTTGTAACTTT
Phenotype
MIM: 608813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DERL1 PubMed Links

Gene Details

Gene
DERL1
Gene Name
derlin 1
There are no transcripts associated with this gene.

Gene
TBC1D31
Gene Name
TBC1 domain family member 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145088.1 Intron NP_001138560.1
NM_145647.3 Intron NP_663622.2
XM_005251102.1 Intron XP_005251159.1
XM_005251103.1 Intron XP_005251160.1
XM_005251104.1 Intron XP_005251161.1
XM_011517378.2 Intron XP_011515680.1
XM_011517379.2 Intron XP_011515681.1
XM_017013983.1 Intron XP_016869472.1
XM_017013984.1 Intron XP_016869473.1
XM_017013985.1 Intron XP_016869474.1
XM_017013986.1 Intron XP_016869475.1
XM_017013987.1 Intron XP_016869476.1
XM_017013988.1 Intron XP_016869477.1
XM_017013989.1 Intron XP_016869478.1
XM_017013990.1 Intron XP_016869479.1

View Full Product Details