Product Details

SNP ID
rs12269230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48436390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATCTGTAGAATTATTTCTCTAT[C/T]ACTGAACTTTTCCTAAGTAAAATGT
Phenotype
MIM: 610585 MIM: 601158
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP22 PubMed Links

Gene Details

Gene
ARHGAP22
Gene Name
Rho GTPase activating protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256024.1 2877 Intron NP_001242953.1
NM_001256025.2 2877 Intron NP_001242954.1
NM_001256026.1 2877 Intron NP_001242955.1
NM_021226.3 2877 Intron NP_067049.2
XM_005270014.3 2877 Intron XP_005270071.1
XM_011540002.1 2877 Intron XP_011538304.1
XM_011540003.1 2877 Intron XP_011538305.1
XM_011540004.1 2877 Intron XP_011538306.1
XM_011540005.1 2877 Intron XP_011538307.1
XM_011540006.1 2877 Intron XP_011538308.1
XM_011540007.2 2877 Intron XP_011538309.1
XM_011540011.2 2877 Intron XP_011538313.1
XM_011540012.2 2877 Intron XP_011538314.1
XM_011540013.2 2877 Intron XP_011538315.1
XM_011540015.2 2877 Intron XP_011538317.1
XM_017016470.1 2877 Intron XP_016871959.1
XM_017016471.1 2877 Intron XP_016871960.1
XM_017016472.1 2877 Intron XP_016871961.1
XM_017016473.1 2877 Intron XP_016871962.1
XM_017016474.1 2877 Intron XP_016871963.1
XM_017016475.1 2877 Intron XP_016871964.1
XM_017016476.1 2877 Intron XP_016871965.1
XM_017016477.1 2877 Intron XP_016871966.1
Gene
MAPK8
Gene Name
mitogen-activated protein kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278547.1 2877 UTR 3 NP_001265476.1
NM_001278548.1 2877 UTR 3 NP_001265477.1
NM_001323302.1 2877 UTR 3 NP_001310231.1
NM_001323320.1 2877 UTR 3 NP_001310249.1
NM_001323321.1 2877 UTR 3 NP_001310250.1
NM_001323322.1 2877 UTR 3 NP_001310251.1
NM_001323323.1 2877 UTR 3 NP_001310252.1
NM_001323324.1 2877 UTR 3 NP_001310253.1
NM_001323325.1 2877 UTR 3 NP_001310254.1
NM_001323326.1 2877 UTR 3 NP_001310255.1
NM_001323327.1 2877 UTR 3 NP_001310256.1
NM_001323328.1 2877 UTR 3 NP_001310257.1
NM_001323329.1 2877 UTR 3 NP_001310258.1
NM_001323330.1 2877 UTR 3 NP_001310259.1
NM_001323331.1 2877 UTR 3 NP_001310260.1
NM_139046.3 2877 UTR 3 NP_620634.1
NM_139049.3 2877 UTR 3 NP_620637.1

View Full Product Details