Product Details

SNP ID
rs10860216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:97474033 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAAGGGAAGAATTGGCTAAAAAA[A/G]TACACATTCCTAGTAGACATTTCTT
Phenotype
MIM: 607045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RMST PubMed Links

Gene Details

Gene
RMST
Gene Name
rhabdomyosarcoma 2 associated transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details