Product Details

SNP ID
rs4653166
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:36238687 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTAGAAGATTTAGTGTAATGAG[C/T]GAGCTTTTCGGGCTTTTGTTTTGTT
Phenotype
MIM: 603809
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THRAP3 PubMed Links

Gene Details

Gene
THRAP3
Gene Name
thyroid hormone receptor associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321471.1 Intron NP_001308400.1
NM_001321473.1 Intron NP_001308402.1
NM_005119.3 Intron NP_005110.2
XM_005271371.4 Intron XP_005271428.1

View Full Product Details