Product Details

SNP ID
rs11264112
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:35179742 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCCCACCCCAATAAAGTTCCTGA[A/C]GTCCATATATTATATACCAAGTACA
Phenotype
MIM: 605199
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 6950 Intron NP_005057.1
XM_005271112.4 6950 Intron XP_005271169.1
XM_005271113.4 6950 Intron XP_005271170.1
XM_005271115.4 6950 Intron XP_005271172.1
XM_011541950.2 6950 UTR 3 XP_011540252.1
XM_017002053.1 6950 UTR 3 XP_016857542.1
XM_017002054.1 6950 UTR 3 XP_016857543.1

View Full Product Details