Product Details

SNP ID
rs13353057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102645688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCAGCGCTATTGGAAACACACT[A/G]GGGGGGGAGTGGGGGGAGTTTCCAG
Phenotype
MIM: 606125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM8 PubMed Links

Gene Details

Gene
TRIM8
Gene Name
tripartite motif containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030912.2 Intron NP_112174.2

View Full Product Details