Product Details

SNP ID
rs7157257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24049930 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGACAAATGCCAGCAATGCCTAA[C/T]TCCGTACTGGTCCAGACATCCCAAA
Phenotype
MIM: 614716 MIM: 615195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARMIL3 PubMed Links

Gene Details

Gene
CARMIL3
Gene Name
capping protein regulator and myosin 1 linker 3
There are no transcripts associated with this gene.

Gene
DHRS4L1
Gene Name
dehydrogenase/reductase 4 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277864.1 Intron NP_001264793.1
XM_006720246.3 Intron XP_006720309.1

View Full Product Details