Product Details

SNP ID
rs12279365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:110181633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGGCATGGCCCCTGTGCCTTC[A/G]TTTTACAATGTGTGGCTTTGAAGGG
Phenotype
MIM: 179410 MIM: 615001
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RDX PubMed Links

Gene Details

Gene
RDX
Gene Name
radixin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260492.1 Intron NP_001247421.1
NM_001260493.1 Intron NP_001247422.1
NM_001260494.1 Intron NP_001247423.1
NM_001260495.1 Intron NP_001247424.1
NM_001260496.1 Intron NP_001247425.1
NM_002906.3 Intron NP_002897.1
Gene
ZC3H12C
Gene Name
zinc finger CCCH-type containing 12C
There are no transcripts associated with this gene.

View Full Product Details