Product Details

SNP ID
rs12796659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14879949 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGCATAATAAGACCTCTTTCAG[A/T]GATTTTTTTTACATGAATTGAGATA
Phenotype
MIM: 608713 MIM: 602047
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP2R1 PubMed Links

Gene Details

Gene
CYP2R1
Gene Name
cytochrome P450 family 2 subfamily R member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024514.4 Intron NP_078790.2
XM_005252788.1 Intron XP_005252845.1
XM_005252789.2 Intron XP_005252846.1
XM_011519895.1 Intron XP_011518197.1
XM_011519898.2 Intron XP_011518200.1
XM_017017190.1 Intron XP_016872679.1
XM_017017191.1 Intron XP_016872680.1
XM_017017192.1 Intron XP_016872681.1
XM_017017193.1 Intron XP_016872682.1
XM_017017194.1 Intron XP_016872683.1
XM_017017195.1 Intron XP_016872684.1
Gene
PDE3B
Gene Name
phosphodiesterase 3B
There are no transcripts associated with this gene.

View Full Product Details