Product Details

SNP ID
rs12720448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223053103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGAAGCCGCTGCCTGTGGTGTC[C/G]GGCCTGAGGTCGGTGCAGGTGCCCC
Phenotype
MIM: 607775
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNE4 PubMed Links

Gene Details

Gene
KCNE4
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080671.3 641 Silent Mutation TCC,TCG S142S NP_542402.3

View Full Product Details