Product Details

SNP ID
rs11634919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55543971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACACTACTGAACACCATCAAATG[C/T]CTGTCAGCCCAAAATACTGCATTTT
Phenotype
MIM: 606902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYGO1 PubMed Links

Gene Details

Gene
PYGO1
Gene Name
pygopus family PHD finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015617.2 3707 UTR 3 NP_056432.1
XM_005254300.4 3707 UTR 3 XP_005254357.1
XM_011521446.2 3707 UTR 3 XP_011519748.1

View Full Product Details