Product Details

SNP ID
rs12924557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:426279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACCCCGGGCCGTCCGCCCCGCC[A/G]CCGCGCTCCGGCCCGCGGGGGCAGC
Phenotype
MIM: 608738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB11FIP3 PubMed Links

Gene Details

Gene
RAB11FIP3
Gene Name
RAB11 family interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142272.1 658 Intron NP_001135744.1
NM_014700.3 658 Silent Mutation CCA,CCG P91P NP_055515.1
XM_005255713.3 658 Silent Mutation CCA,CCG P91P XP_005255770.1
XM_005255714.1 658 Intron XP_005255771.1
XM_005255715.4 658 Intron XP_005255772.1
XM_005255717.3 658 Silent Mutation CCA,CCG P91P XP_005255774.1
XM_005255718.3 658 Silent Mutation CCA,CCG P91P XP_005255775.1
XM_011522764.2 658 Intron XP_011521066.1
XM_011522765.2 658 Intron XP_011521067.1
XM_017023907.1 658 Silent Mutation CCA,CCG P91P XP_016879396.1

View Full Product Details