Product Details

SNP ID
rs11980069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30597452 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTTTTAAAATTATTTTTCCTTT[C/T]CCACCTACATATGTCTTTCTTCATA
Phenotype
MIM: 600287
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GARS PubMed Links

Gene Details

Gene
GARS
Gene Name
glycyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316772.1 Intron NP_001303701.1
NM_002047.3 Intron NP_002038.2
XM_006715686.2 Intron XP_006715749.1

View Full Product Details