Product Details

SNP ID
rs11800087
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:17374076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACACCCCCTCTACCAATTAAAC[C/T]TGAGGTACCAATGGAACCTCAGGGC
Phenotype
MIM: 610363
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PADI6 PubMed Links

Gene Details

Gene
PADI6
Gene Name
peptidyl arginine deiminase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207421.4 Intron NP_997304.3

View Full Product Details