Product Details

SNP ID
rs7957464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9951690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCTTGGGACTTGCTTATTGTTG[C/T]AGCAGTTAGATTGTATTCTGGTGGT
Phenotype
MIM: 612088
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLEC12A PubMed Links

Gene Details

Gene
CLEC12A
Gene Name
C-type lectin domain family 12 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207010.1 Intron NP_001193939.1
NM_001300730.1 Intron NP_001287659.1
NM_138337.5 Intron NP_612210.4
NM_201623.3 Intron NP_963917.2
XM_005253324.2 Intron XP_005253381.1
XM_006719033.3 Intron XP_006719096.1
XM_006719035.3 Intron XP_006719098.1
XM_006719036.3 Intron XP_006719099.1
XM_011520570.1 Intron XP_011518872.1
XM_011520571.2 Intron XP_011518873.2
XM_011520573.2 Intron XP_011518875.1
Gene
LOC100506159
Gene Name
uncharacterized LOC100506159
There are no transcripts associated with this gene.

Gene
LOC400002
Gene Name
uncharacterized LOC400002
There are no transcripts associated with this gene.

View Full Product Details