Product Details

SNP ID
rs11968543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32046114 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTGCAGTCATCTTTGTCTTCAG[A/C]CCAAATGCACAAGGAAACCCACACA
Phenotype
MIM: 613815 MIM: 600985
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYP21A2 PubMed Links

Gene Details

Gene
CYP21A2
Gene Name
cytochrome P450 family 21 subfamily A member 2
There are no transcripts associated with this gene.

Gene
TNXB
Gene Name
tenascin XB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019105.6 15 Intron NP_061978.6
NM_032470.3 15 UTR 5 NP_115859.2

View Full Product Details