Product Details

SNP ID
rs1816702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153688371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAACTTAGAATTACAATGGACTGC[C/T]GAGACCAGCTCGGTCGTGGAGACCC
Phenotype
MIM: 603028
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100419170 PubMed Links

Gene Details

Gene
LOC100419170
Gene Name
toll like receptor 2 pseudogene
There are no transcripts associated with this gene.

Gene
TLR2
Gene Name
toll like receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318787.1 Intron NP_001305716.1
NM_001318789.1 Intron NP_001305718.1
NM_001318790.1 Intron NP_001305719.1
NM_001318791.1 Intron NP_001305720.1
NM_001318793.1 Intron NP_001305722.1
NM_001318795.1 Intron NP_001305724.1
NM_001318796.1 Intron NP_001305725.1
NM_003264.4 Intron NP_003255.2
XM_011532215.2 Intron XP_011530517.1
XM_011532216.2 Intron XP_011530518.1
XM_017008573.1 Intron XP_016864062.1
XM_017008574.1 Intron XP_016864063.1
XM_017008575.1 Intron XP_016864064.1
XM_017008576.1 Intron XP_016864065.1

View Full Product Details