Product Details

SNP ID
rs13190731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137870943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGACAAACATTACTGAAAAC[A/C]CGGGTTATTCCCCTCCTCCTCAGTA
Phenotype
MIM: 191163
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130476 PubMed Links

Gene Details

Gene
LOC100130476
Gene Name
uncharacterized LOC100130476
There are no transcripts associated with this gene.

Gene
LOC107986649
Gene Name
uncharacterized LOC107986649
There are no transcripts associated with this gene.

Gene
TNFAIP3
Gene Name
TNF alpha induced protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270507.1 Intron NP_001257436.1
NM_001270508.1 Intron NP_001257437.1
NM_006290.3 Intron NP_006281.1
XM_005267119.1 Intron XP_005267176.1
XM_006715555.1 Intron XP_006715618.1
XM_011536095.1 Intron XP_011534397.1
XM_011536096.1 Intron XP_011534398.1

View Full Product Details