Product Details

SNP ID
rs13208524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132745523 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTATCAAACTCTGCCAACAGAT[C/G]CATTCTTGAAGCTGGTGGGATCATG
Phenotype
MIM: 603571
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VNN2 PubMed Links

Gene Details

Gene
VNN2
Gene Name
vanin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242350.2 Intron NP_001229279.1
NM_004665.4 Intron NP_004656.2
NM_078488.2 Intron NP_511043.1
XM_006715593.3 Intron XP_006715656.1
XM_011536231.1 Intron XP_011534533.1
XM_017011407.1 Intron XP_016866896.1
XM_017011408.1 Intron XP_016866897.1
XM_017011409.1 Intron XP_016866898.1

View Full Product Details