Product Details

SNP ID
rs12329829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:28575864 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCACATAGTATGTTCTCCGTGTTT[A/G]TTCTTTCACTCAACATAAAGTTTCT
Phenotype
MIM: 614553
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
N6AMT1 PubMed Links

Gene Details

Gene
N6AMT1
Gene Name
N-6 adenine-specific DNA methyltransferase 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013240.5 Intron NP_037372.4
NM_182749.4 Intron NP_877426.4

View Full Product Details