Product Details

SNP ID
rs12989017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:173358744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCAGAGTAAAGAAGAACTTAAA[C/G]AAATTCAGATATGTGAAGTTGATTT
Phenotype
MIM: 609937
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDCA7 PubMed Links

Gene Details

Gene
CDCA7
Gene Name
cell division cycle associated 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031942.4 185 Missense Mutation AAC,AAG N18K NP_114148.3
NM_145810.2 185 Missense Mutation AAC,AAG N18K NP_665809.1
XM_011511957.2 185 UTR 5 XP_011510259.1

View Full Product Details