Product Details

SNP ID
rs139462088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47737815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAAGGGGCGCGCGGGGCCGCC[G/T]GGAAGAAGCTCCAGGGGCCCCGCGC
Phenotype
MIM: 116898 MIM: 615384
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEBPD PubMed Links

Gene Details

Gene
CEBPD
Gene Name
CCAAT/enhancer binding protein delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005195.3 350 Silent Mutation NP_005186.2
Gene
SPIDR
Gene Name
scaffolding protein involved in DNA repair
There are no transcripts associated with this gene.

View Full Product Details