Product Details

SNP ID
rs3957094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73114426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTTGGTGCTATAGGCTGCTG[C/T]CAACCTATGTTACTAAGACACGGTG
Phenotype
MIM: 609233
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT13 PubMed Links

Gene Details

Gene
NUDT13
Gene Name
nudix hydrolase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283014.1 263 Missense Mutation CCA,TCA P21S NP_001269943.1
NM_001283015.1 263 Missense Mutation CCA,TCA P21S NP_001269944.1
NM_001283016.1 263 UTR 5 NP_001269945.1
NM_001283017.1 263 UTR 5 NP_001269946.1
NM_001283019.1 263 UTR 5 NP_001269948.1
NM_015901.5 263 Missense Mutation CCA,TCA P21S NP_056985.3
XM_017016033.1 263 Missense Mutation CCA,TCA P21S XP_016871522.1

View Full Product Details