Product Details

SNP ID
rs3172495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48599681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGTCATAGATGTACTTGGAGCA[A/G]ATCTCACGTACCACACTGGCATCCA
Phenotype
MIM: 120120 MIM: 191328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL7A1 PubMed Links

Gene Details

Gene
COL7A1
Gene Name
collagen type VII alpha 1
There are no transcripts associated with this gene.

Gene
SNORA94
Gene Name
small nucleolar RNA, H/ACA box 94
There are no transcripts associated with this gene.

Gene
UQCRC1
Gene Name
ubiquinol-cytochrome c reductase core protein I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003365.2 1377 Silent Mutation ATC,ATT I444I NP_003356.2

View Full Product Details