Product Details
- SNP ID
-
rs3783497
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2476787 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTCTGCCGTTTTGTGGATCGGGG[A/G]CTGCCGTATCCTGATGTATCGTCTG
- Phenotype
-
MIM: 604948
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GADD45B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3783498] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GADD45B
- Gene Name
- growth arrest and DNA damage inducible beta
View Full Product Details