Product Details

SNP ID
rs3176859
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:100721314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACACATTTAGGATTTTTTTTTG[G/T]TTTTTTTGGTGCCATGAAGCCTTGG
Phenotype
MIM: 192225
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VCAM1 PubMed Links

Gene Details

Gene
VCAM1
Gene Name
vascular cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078.3 Intron NP_001069.1
NM_001199834.1 Intron NP_001186763.1
NM_080682.2 Intron NP_542413.1

View Full Product Details