Product Details

SNP ID
rs4477177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:14881290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAATGCCTTCATTTTGTTAGG[C/G]TGAAGGAGACAAAGTGGTTAGAGAA
Phenotype
MIM: 300515
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FANCB PubMed Links

Gene Details

Gene
FANCB
Gene Name
Fanconi anemia complementation group B
There are no transcripts associated with this gene.

Gene
MOSPD2
Gene Name
motile sperm domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177475.1 Intron NP_001170946.1
NM_152581.3 Intron NP_689794.1

View Full Product Details